Order Entry
Austria
ContactUsLinkComponent
1,3-Dimethyl-5-(hydroxymethyl)-1H-pyrazole
Irritant
1,3-Dimethyl-5-(hydroxymethyl)-1H-pyrazole
Catalog # APOSOR23188-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
1,3-Dimethyl-5-(hydroxymethyl)-1H-pyrazole
Catalog # APOSOR23188-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :126.156 g/mol
Boiling Pt:117…118 °C (0.8 mm Hg)
 :GHS07
Storage Temperature:Ambient
CAS Number:57012-20-1
Formula:C₆H₁₀N₂O
MDL Number:MFCD02682037