
N-(2-Chloroethyl)-1H-pyrazole-1-carboxamide
Catalog # APOSOR16157-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:173.6 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:1209343-75-8
Formula:C₆H₈ClN₃O
MDL Number:MFCD11109511