Ethyl 4-bromo-5-methyl-1H-pyrazole-3-carboxylate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:6076-14-8
:
Formula:C₇H₉BrN₂O₂
MDL Number:MFCD11054303
Melting Pt:131…133 °C
:233.063 g/mol
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together