Ethyl 1-(3,5-dichlorophenyl)-3,5-dimethyl-1H-pyrazole-4-carboxylate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1858255-30-7
:
Formula:C₁₄H₁₄Cl₂N₂O₂
MDL Number:MFCD28347986
:313.179 g/mol
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together