[4-(4-Bromo-1H-pyrazol-1-yl)phenyl]methanol
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1184193-54-1
:
Formula:C₁₀H₉BrN₂O
MDL Number:MFCD12756251
:253.095 g/mol
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together