Ethyl 1-(4-bromophenyl)-3,5-dimethyl-1H-pyrazole-4-carboxylate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:113808-82-5
:
Formula:C₁₄H₁₅BrN₂O₂
MDL Number:MFCD11973596
:323.185 g/mol
Storage Temperature:Ambient
Frequently Bought Together