Order Entry
Austria
ContactUsLinkComponent
1,3-Dimethyl-1H-pyrazole-5-carboxamide
Irritant
1,3-Dimethyl-1H-pyrazole-5-carboxamide
Catalog # APOSOR0420-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
1,3-Dimethyl-1H-pyrazole-5-carboxamide
Catalog # APOSOR0420-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :139.155 g/mol
Melting Pt:165 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:136678-93-8
Formula:C₆H₉N₃O
MDL Number:MFCD00833259