
Silicon dioxide 99,999%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:14808-60-7
:7631-86-9
Formula:SiO₂
MDL Number:MFCD00011232
:60.08 g/mol
Density:2.6 g/cm³ (25 °C)
:GHS07
Storage Temperature:Ambient
Frequently Bought Together