Order Entry
Austria
ContactUsLinkComponent
 
t-Boc-N-Amido-PEG2-CH2CO2H
Catalog # APOSBIPG1825-1G
Supplier:  Apollo Scientific
CAS Number:  
t-Boc-N-Amido-PEG2-CH2CO2H
Catalog # APOSBIPG1825-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Boc-NH-PEG2-CH2COOH, 2,2-Dimethyl-4-oxo-3,8,11-trioxa-5-azatridecan-13-oic acid
 :
 :263.288 g/mol
Storage Temperature:Ambient
CAS Number:108466-89-3
Formula:C₁₁H₂₁NO₆
MDL Number:MFCD17019374