Order Entry
Austria
ContactUsLinkComponent
Sulpho-SAED (Sulphosuccinimidyl 2-(7-azido-4-methylcoumarin-3-acetamide)ethyl-1,3-dithiopropionate)
Sulpho-SAED (Sulphosuccinimidyl 2-(7-azido-4-methylcoumarin-3-acetamide)ethyl-1,3-dithiopropionate)
Catalog # APOSBIPA113-50MG
Supplier:  Apollo Scientific
CAS Number:  
Sulpho-SAED (Sulphosuccinimidyl 2-(7-azido-4-methylcoumarin-3-acetamide)ethyl-1,3-dithiopropionate)
Catalog # APOSBIPA113-50MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    50 mg
  • Seq:
    2370
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Sulfo SAED
 :
 :671.677 g/mol
 :GHS07
Storage Temperature:Ambient
Formula:C₂₄H₂₅N₅O₁₂S₃
MDL Number:MFCD01861966