
5-Chloro-8-hydroxyquinoline β-D-glucuronide ≥98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:65851-39-0
:
Formula:C₁₅H₁₄ClNO₇
MDL Number:MFCD29924274
:355.727 g/mol
Melting Pt:206…208 °C
:GHS07
Storage Temperature:Freezer
Frequently Bought Together