
5,7-Dichloro-8-hydroxyquinoline β-D-glucuronide ≥98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:40951-47-1
:
Formula:C₁₅H₁₃Cl₂NO₇
MDL Number:MFCD29924273
:390.172 g/mol
Melting Pt:175…178 °C
:GHS07
Storage Temperature:Freezer
Frequently Bought Together