1,3,4,6-Tetra-O-acetyl-2-azido-2-deoxy-D-galactopyranose ≥98%
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: 2-Azido-2-deoxy-D-galactose peracetate, 1,3,4,6-Tetra-O-acetyl-2-azido-2-deoxy-D-galactopyranose, 2-Azido-2-deoxy-D-galactopyranose 1,3,4,6-tetraacetate
CAS Number:84278-00-2
:
Formula:C₁₄H₁₉N₃O₉
MDL Number:MFCD02683262
:373.315 g/mol
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together