1,6-Anhydro-2-azido-2-deoxy-β-D-glucopyranose ≥99%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:67546-20-7
:
Formula:C₆H₉N₃O₄
MDL Number:MFCD16660958
Melting Pt:128…129 °C
:187.153 g/mol
:GHS07
Storage Temperature:Freezer
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together