
5-Cyano-2,3-ditolyl tetrazolium chloride
Catalog # APOSBIC1455-50MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:50 mg
- Seq:2370
Specifications
About this item
:
:311.769 g/mol
Melting Pt:255 °C
:GHS07
Storage Temperature:Refrigerator
CAS Number:90217-02-0
Formula:C₁₆H₁₄N₅·Cl
MDL Number:MFCD00467521