Order Entry
Austria
Orders LinkContactUsLinkComponent
Cochlioquinone A

Cochlioquinone A

Irritant
Supplier:   Apollo Scientific
Cochlioquinone A
Supplier:  Apollo Scientific
Certificates
Cochlioquinone A

2 Options Available Below

Product Details & Documents

From microbial source. Purity: (c.70%)

Synonyms:  Luteoleersin
  • CAS Number:32450-25-2
  • Formula:C₃₀H₄₄O₈
  • MDL Number:MFCD08062299
  • Molecular Weight:532.666 g/mol
  • Storage Temperature:Freezer
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    1 mg
    Log in for availability
    Log in for availability
    1 * 1 mg
    1 * 1 mg - 174,00 €
    174,00 €
    1 * 1 mg
       174,00 €

  • Bottle
    5 mg
    Log in for availability
    Log in for availability
    1 * 5 mg
    1 * 5 mg - 710,00 €
    710,00 €
    1 * 5 mg
       710,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.