N,O-Diacetyltyramine
Catalog # APOSBIBR1141-1MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 mg
- Seq:2070
Specifications
About this item
:
:221.252 g/mol
Storage Temperature:Ambient
CAS Number:14383-56-3
Formula:C₁₂H₁₅NO₃
MDL Number:MFCD00179163