Order Entry
Austria
ContactUsLinkComponent
 
Irritant
Griseoviridin
Catalog # APOSBIBR1137-1MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Griseoviridin
Catalog # APOSBIBR1137-1MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 mg
  • Seq:
    2070

Specifications

About this item

 :
 :477.531 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:53216-90-3
Formula:C₂₂H₂₇N₃O₇S
MDL Number:MFCD01724491