ContactUsLinkComponent
Cleanroom overalls, hooded, ProShield® 60
Cleanroom overalls, hooded, ProShield® 60
Supplier:  DuPont
Certificates
undefined
Cleanroom overalls, hooded, ProShield® 60
Supplier:  DuPont
Specifications
  • Catalog No:
  • DUPOP6127SWH00/7XL
  • DUPOP6127SWH00/4XL
  • DUPOP6127SWH00/L
  • DUPOP6127SWH00/3XL
  • DUPOP6127SWH00/M
  • DUPOP6127SWH00/6XL
  • DUPOP6127SWH00/2XL
  • DUPOP6127SWH00/XL
  • DUPOP6127SWH00/S
  • DUPOP6127SWH00/5XL
  • Model:
  • P6127S
  • P6127S
  • P6127S
  • P6127S
  • P6127S
  • P6127S
  • P6127S
  • P6127S
  • P6127S
  • P6127S
  • Fabric:
  • PP
  • PP
  • PP
  • PP
  • PP
  • PP
  • PP
  • PP
  • PP
  • PP
  • Closure type:
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Anti-static:
  • +
  • +
  • +
  • +
  • +
  • +
  • +
  • +
  • +
  • +
  • Gender:
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Garment application:
  • General Purpose, Laboratory, Industrial
  • General Purpose, Laboratory, Industrial
  • General Purpose, Laboratory, Industrial
  • General Purpose, Laboratory, Industrial
  • General Purpose, Laboratory, Industrial
  • General Purpose, Laboratory, Industrial
  • General Purpose, Laboratory, Industrial
  • General Purpose, Laboratory, Industrial
  • General Purpose, Laboratory, Industrial
  • General Purpose, Laboratory, Industrial
  • Tensile strength:
  • >30 N
  • >30 N
  • >30 N
  • >30 N
  • >30 N
  • >30 N
  • >30 N
  • >30 N
  • >30 N
  • >30 N
  • Weight:
  • 60 g/m²
  • 60 g/m²
  • 60 g/m²
  • 60 g/m²
  • 60 g/m²
  • 60 g/m²
  • 60 g/m²
  • 60 g/m²
  • 60 g/m²
  • 60 g/m²