ContactUsLinkComponent
Cleanroom boot covers, Tyvek® IsoClean®, model IC451S, non sterile
Cleanroom boot covers, Tyvek® IsoClean®, model IC451S, non sterile
Supplier:  DuPont
Certificates
undefined
Cleanroom boot covers, Tyvek® IsoClean®, model IC451S, non sterile
Supplier:  DuPont
Specifications
  • Catalog No:
  • DUPOIC451SWH00/L
  • DUPOIC451SWH00/M
  • Fabric:
  • Tyvek®
  • Tyvek®
  • Gender:
  • Unisex
  • Unisex
  • Seam type:
  • Serged
  • Serged
  • Garment application:
  • Cleanroom
  • Cleanroom
  • Tensile strength:
  • >30 N
  • >30 N