Cleanroom frocks, Tyvek® IsoClean®, IC270B, Bulk
Supplier: DuPont
Certificates
About this item
These frocks deliver an ideal balance of protection, durability and comfort. Made using a patented flash spinning process, Tyvek® provides an inherent barrier to particles, microorganisms and non hazardous light liquid splash.
- Bound seams are covered with garment fabric to reinforce the seam and to reduce the potential for particle penetration
- Elastic openings for tighter fit at wrist
- Bound neck for lower particle shedding
- Adjustable snap at neck for better fit
- Body-length jacket design with front snap closures for easy donning and doffing
- Clean-processed garments offer lowest level of particle shedding within DuPont product portfolio
Suitable for use in Class 100, ISO 5, GMP class A/B cleanroom environments.
Length: 99 to 114 cm
Weight: 45 g/m²
Packaging: Packaged in double transparent poly liners. 30 per case. Subgrouped by five in an outer bag.
Specifications
- Catalog No:
- 114-9437
- 114-9436
- 114-9440
- 114-9435
- 114-9438
- 114-9439
- Closure type:
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Cleanroom class:
- 100/ISO 5; EU GMP (A/B)
- 100/ISO 5; EU GMP (A/B)
- 100/ISO 5; EU GMP (A/B)
- 100/ISO 5; EU GMP (A/B)
- 100/ISO 5; EU GMP (A/B)
- 100/ISO 5; EU GMP (A/B)
- Pockets:
- Without pockets
- Without pockets
- Without pockets
- Without pockets
- Without pockets
- Without pockets
- Gender:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Seam type:
- Bound
- Bound
- Bound
- Bound
- Bound
- Bound
- Garment application:
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Water resistance:
- 7 kPa
- 7 kPa
- 7 kPa
- 7 kPa
- 7 kPa
- 7 kPa
- Thickness:
- 185 µm
- 185 µm
- 185 µm
- 185 µm
- 185 µm
- 185 µm
- Weight:
- 45 g/m²
- 45 g/m²
- 45 g/m²
- 45 g/m²
- 45 g/m²
- 45 g/m²
Specifications
Frequently Bought Together