

Hydrochloric acid 37% (max. 0.005 ppm Hg), AnalaR NORMAPUR® analytical reagent
Catalog # 88372.320
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:2,5 L
- Seq:5096
Specifications
Specification Test Results
- Assay:37 - 38%
- Ammonium (NH4):max. 3 ppm
- Free Chlorine:max. 0,5 ppm
- Mercury (Hg):max. 5 ppb
- Residue after ignition:max. 5 ppm
- Sulphate (SO4):max. 1 ppm
- Sulfite (SO3):max. 2 ppm
- Aluminium (Al):max. 0,05 ppm
- Arsenic (As):max. 0,05 ppm
- Barium (Ba):max. 0,02 ppm
- Beryllium (Be):max. 0,01 ppm
- Cadmium (Cd):max. 0,02 ppm
- Calcium (Ca):max. 0,5 ppm
- Chromium (Cr):max. 0,05 ppm
- Cobalt (Co):max. 0,01 ppm
- Copper (Cu):max. 0,01 ppm
- Germanium (Ge):max. 0,1 ppm
- Iron (Fe):max. 0,2 ppm
- Lead (Pb):max. 0,05 ppm
- Lithium (Li):max. 0,01 ppm
- Magnesium (Mg):max. 0,05 ppm
- Manganese (Mn):max. 0,01 ppm
- Molybdenum (Mo):max. 0,02 ppm
- Nickel (Ni):max. 0,05 ppm
- Potassium (K):max. 0,1 ppm
- Silver (Ag):max. 0,01 ppm
- Sodium (Na):max. 0,3 ppm
- Strontium (Sr):max. 0,01 ppm
- Thallium (Tl):max. 0,05 ppm
- Titanium (Ti):max. 0,05 ppm
- Vanadium (V):max. 0,01 ppm
- Zinc (Zn):max. 0,5 ppm
- Zirconium (Zr):max. 0,05 ppm
Specification Test Results
About this item
:
ADR:8,II
:36.46 g/mol
UN:1789
:GHS05
:GHS07
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324