Order Entry
Austria
ContactUsLinkComponent
 
 
 
 
Nitric acid 65% (max. 0.005 ppm Hg), AnalaR NORMAPUR® analytical reagent
 
undefined
 
 
 
Nitric acid 65% (max. 0.005 ppm Hg), AnalaR NORMAPUR® analytical reagent
 

 

  • Assay:
    min. 65%
  • Chloride (Cl):
    max. 0,5 ppm
  • Dithizone determination:
    passes test
  • Mercury (Hg):
    max. 5 ppb
  • Residue after ignition:
    max. 5 ppm
  • Sulphate (SO4):
    max. 1 ppm
  • Aluminium (Al):
    max. 0,05 ppm
  • Arsenic (As):
    max. 0,05 ppm
  • Barium (Ba):
    max. 0,02 ppm
  • Beryllium (Be):
    max. 0,01 ppm
  • Cadmium (Cd):
    max. 0,02 ppm
  • Calcium (Ca):
    max. 0,5 ppm
  • Chromium (Cr):
    max. 0,1 ppm
  • Cobalt (Co):
    max. 0,01 ppm
  • Copper (Cu):
    max. 0,01 ppm
  • Germanium (Ge):
    max. 0,05 ppm
  • Iron (Fe):
    max. 0,1 ppm
  • Lead (Pb):
    max. 0,05 ppm
  • Lithium (Li):
    max. 0,01 ppm
  • Magnesium (Mg):
    max. 0,1 ppm
  • Manganese (Mn):
    max. 0,01 ppm
  • Molybdenum (Mo):
    max. 0,02 ppm
  • Nickel (Ni):
    max. 0,05 ppm
  • Potassium (K):
    max. 0,1 ppm
  • Silver (Ag):
    max. 0,01 ppm
  • Sodium (Na):
    max. 0,5 ppm
  • Strontium (Sr):
    max. 0,01 ppm
  • Thallium (Tl):
    max. 0,05 ppm
  • Titanium (Ti):
    max. 0,1 ppm
  • Vanadium (V):
    max. 0,01 ppm
  • Zinc (Zn):
    max. 0,05 ppm
  • Zirconium (Zr):
    max. 0,1 ppm

 

Frequently Bought Together