Order Entry
Austria
ContactUsLinkComponent
 
VWR® pH 130, pH/mV Meter, Portable
Catalog # 662-2351
Supplier:  VWR Collection
undefined
VWR® pH 130, pH/mV Meter, Portable
Catalog # 662-2351
Supplier:  VWR Collection

Specifications

  • Temperature range:
    0,0...50,0 °C
  • Description:
    Portable pH/mV meter set, pH 130, with touch screen
  • Temperature accuracy:
    ±0,1 °C
  • Temperature resolution:
    0,1 °C
  • Input:
    10¹² Ω
  • Dimensions:
    149×82×22 mm
  • Power supply:
    2× AA 1,2 V rechargeable battery, 5 V/1000 mA power adapter
  • Temperature compensation:
    –5...+110,0 °C
  • Weight:
    0,24 kg
  • mV accuracy:
    ±1 mV
  • mV range:
    ±2000 mV
  • mV resolution:
    1 mV
  • pH accuracy:
    ±0,01 pH
  • pH range:
    0,00 - 14,00
  • pH resolution:
    0,01 pH
  • Pk:
    1

Specifications

About this item

These economical, robust, and waterproof pH meters are supplied in complete, ready to use kits that incorporate a robust carrying case, electrode and temperature sensor with different cable lengths and buffers.

  • Colour, graphic touch screen - enables simultaneous read-out of the pH or mV and temperature value
  • Waterproof housing (IP 66) enables work in difficult conditions
  • Small size and low weight make working in the field easier
  • Calibration of the pH electrode: 1 to 3 points (pH: 4,00 / 7,00 / 10,00)
  • Automatic buffer detection; calibration time reminder
  • Automatic temperature compensation
  • Read-out of the electrode's slope and offset; memory of 200 results

Used for pH, mV (redox potential) and temperature measurements. Powered by rechargeable batteries or power adapter. Long lasting work in laboratory possible with use of the power adapter.

Ordering information: 24 month warranty.
Delivery: Equipped with a standard pH electrode with built-in temperature sensor.

Buffer solutions, accessory, 20 °C AVS TITRINORM

Product Information