Order Entry
Austria
ContactUsLinkComponent
 
VWR®, D.F.B (Decalcifying solution - H.K. Kristenson) Histological grade
Catalog # 352432C
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR®, D.F.B (Decalcifying solution - H.K. Kristenson) Histological grade
Catalog # 352432C
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bag
  • Pk:
    5 L
  • Description:
    D.F.B. (decalcifying solution - H K Kristensons)

Specifications

About this item

Decalcifier according to Kristenson.

  • pH (25°C) 2.20 --> 2.70