ContactUsLinkComponent
VWR® VINYL HYBRID, Gloves, Vinyl
VWR® VINYL HYBRID, Gloves, Vinyl
Supplier:  VWR Collection
Certificates
undefined
VWR® VINYL HYBRID, Gloves, Vinyl
Supplier:  VWR Collection
Specifications
  • Catalog No:
  • 112-6072
  • 112-6073
  • Ambidextrous / hand-specific:
  • Ambidextrous
  • Ambidextrous
  • Disposable / reusable:
  • Disposable
  • Disposable
  • Latex / latex-free:
  • Latex-free
  • Latex-free
  • Powdered / powder-free:
  • Powder-free
  • Powder-free
  • Finish:
  • Smooth
  • Smooth
  • Cuff style:
  • Beaded
  • Beaded
  • Finger thickness:
  • 0,18 mm
  • 0,18 mm
  • Palm thickness:
  • 0,16 mm
  • 0,16 mm
  • EN norm:
  • EN ISO 374-1 Type B (KTP), EN 374-5 VIRUS
  • EN ISO 374-1 Type B (KTP), EN 374-5 VIRUS
  • Food contact approved:
  • +
  • +