Order Entry
Austria
ContactUsLinkComponent
 
Formaldehyde 4% in aqueous solution, VOLUSOL®, buffered pH 7.2 -7.6
Catalog # 6368.5000
Supplier:  VWR Chemicals
CAS Number:  
undefined
Formaldehyde 4% in aqueous solution, VOLUSOL®, buffered pH 7.2 -7.6
Catalog # 6368.5000
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pk:
    5 L
  • Seq:
    5190
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Assay (Formaldehyde):
    3.5 --> 4.5 %
  • pH (20°C):
    7.2 --> 7.6
  • Methanol:
    0.5 --> 1.5 %

Specification Test Results

About this item

Synonyms: Paraformaldehyde aqueous solution, Methanal solution, Methylaldehyde solution, Formol, Formaline solution, Formalin, Methylaldehyde
 :
 :30.03 g/mol
 :Stabilised with methanol 0,5-1,5 %
EINECS:200-001-8
 :GHS08
 :GHS07
Storage Temperature:Ambient
CAS Number:50-00-0
Formula:CH₂O
MDL Number:MFCD00003274