ContactUsLinkComponent
Cone folded filter papers, Whatman™
Cone folded filter papers, Whatman™
Certificates
undefined
Cone folded filter papers, Whatman™
Specifications
  • Catalog No:
  • WHAT1002-10240
  • WHAT990010112
  • WHAT990010116
  • WHAT990110116
  • Grade:
  • 2
  • 40
  • 40
  • 1
  • Thickness:
  • 210 µm
  • 210 µm
  • 180 µm
  • Ash content:
  • 0,007%
  • 0,007%
  • 0,06%
  • Basis weight:
  • 95 g/m²
  • 95 g/m²
  • 87 g/m²