


Wright's stain solution for microscopy, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Certificates
About this item
CAS Number:68988-92-1
:51938-72-8
MDL Number:MFCD00005038
Density:0,80 g/ml (20 °C)
ADR:3,II
:M-Clarity™ quality level = MQ100
Flash Pt:11 °C
UN:1230
:GHS02
:GHS06
:GHS08
Specification Test Results
- Appearance (colour):Blue to very dark blue
- Appearance (form):Liquid
- Thin layer chromatogr.:Corresponds to standard
- Density D20/4:0.785 - 0.814
- Wavelength (1) (UV):635 - 650 nm
- Extinction (1%/1 cm) (1) (UV):--
- Wavelength (2) (UV):512 - 527 nm
- Extinction (1%/1 cm) (2) (UV):--
- Solvent (UV):Meoh
- Expiry period:30 months
Specification Test Results
Frequently Bought Together