Order Entry
Austria
ContactUsLinkComponent
4,4′-Dinitro-2-biphenylamine ≥98%, Sigma-Aldrich®
Irritant
4,4′-Dinitro-2-biphenylamine ≥98%, Sigma-Aldrich®
Catalog # 335428-25G
CAS Number:  
undefined
Irritant
4,4′-Dinitro-2-biphenylamine ≥98%, Sigma-Aldrich®
Catalog # 335428-25G
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :259.22 g/mol
Melting Pt:208…210 °C (lit.)
 :GHS07
Storage Temperature:Ambient
CAS Number:51787-75-8
Formula:C₁₂H₉N₃O₄
MDL Number:MFCD00075056