


Bromocresol green-methyl red solution mixed indicator, Sigma-Aldrich®
Catalog # 318728-500ML
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pk:500 mL
- Seq:2725
Specifications
About this item
:
Density:0.839 g/mL (25 °C)
ADR:3,II
:GHS02
:GHS06
:GHS08
Storage Temperature:Ambient
MDL Number:MFCD00134126