
4,5-Dibromobenzene-1,2-diol 90%, Technical Grade, Sigma-Aldrich®
Catalog # 550248-5G
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance (Colour); White to Grey:Conforms to Requirements
- Appearance (Form); Powder, Crystals, Crystalline Powder and/or Chunks:Conforms to Requirements
- Infrared spectrum:Conforms to Structure
- Proton NMR spectrum:Conforms to Structure
- Carbon:23.7 - 30.1 %
- Purity (GC):≥88.0 %
Specification Test Results
About this item
:
:267.9 g/mol
Melting Pt:111...116 °C (lit.)
:GHS07
Storage Temperature:Room Temperature
CAS Number:2563-26-0
Formula:C₆H₄Br₂O₂
MDL Number:MFCD00233157