Order Entry
Austria
ContactUsLinkComponent
 
Lugol's iodine solution for microscopy, Sigma-Aldrich®
 
undefined
Lugol's iodine solution for microscopy, Sigma-Aldrich®
 

 

  • Appearance (colour):
    Red to very dark red and red-brown and very dark red-brown
  • Appearance (form):
    Liquid
  • Density D20/4:
    1.005 - 1.009
  • Application test:
    Corresponds to requirements

 

Frequently Bought Together