Ethyl 4′-hydroxy-4-biphenylcarboxylate ≥98%, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Certificates
About this item
CAS Number:50670-76-3
:
Formula:C₁₅H₁₄O₃
MDL Number:MFCD07809563
:242.28 g/mol
Melting Pt:142…144 °C (dec., lit.)
:M-Clarity™ quality level = MQ200
:GHS07
Storage Temperature:Room Temperature
Specification Test Results
- Appearance (Colour):White to Yellow
- Appearance (Form):Conforms to Requirements
Powder or Crystals - Infrared spectrum:Conforms to Structure
- Carbon:72.5 - 76.2 %
- Purity (HPLC):≥97.5 %
Specification Test Results
Frequently Bought Together