Order Entry
Austria
ContactUsLinkComponent
Methylene blue solution for microbiology, Sigma-Aldrich®
Methylene blue solution for microbiology, Sigma-Aldrich®
Catalog # 50484-100ML-F
CAS Number:  
undefined
Methylene blue solution for microbiology, Sigma-Aldrich®
Catalog # 50484-100ML-F
CAS Number:  

Specifications

  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :319.86 g/mol
CAS Number:61-73-4
 :MFCD00012111
Formula:C₁₆H₁₈ClN₃S
MDL Number:MFCD00150008