Morin hydrate 1-2 mol/mol water for the determination of Al, Be, Zn, Ga, In, Sc, for microscopy, Sigma-Aldrich®
:654055-01-3
:
:C₁₅H₁₀O₇
:MFCD00217054
:302.24 g/mol
:299...300 °C
:M-Clarity™ quality level = MQ200
:207-542-9
:GHS07
:Room Temperature
- Appearance (Colour):Faint Brown to Brown and Faint Brown-Yellow to Brown Yellow
- Appearance (Form):Powder
- Purity (HPLC):≥85%
- Thin Layer Chromatography:Corresponds to Standard
- Solubility (Colour):Brown to Very Dark Brown and Red to Very Dark Red
- Water:3 - 11 %
- Sulphated Ash:≤3%
- Infrared Spectrum:Conforms to Structure
- Wavelength (1) (UV):360 - 375 nm
- Extinction (1) (UV) (1%/1 cm):≥300
- Solvent (UV):Methanol
Frequently Bought Together