Order Entry
Austria
ContactUsLinkComponent
Sudan IV, Sigma-Aldrich®
Sudan IV, Sigma-Aldrich®
Catalog # 198102-25G
CAS Number:  
undefined
Sudan IV, Sigma-Aldrich®
Catalog # 198102-25G
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :380.45 g/mol
Melting Pt:199 °C (dec., lit.)
Storage Temperature:Ambient
CAS Number:85-83-6
Formula:C₂₄H₂₀N₄O
MDL Number:MFCD00003893