
Diethyl-3,4-pyridinedicarboxylate ≥97%, Sigma-Aldrich®
Catalog # 310212-5G
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:223.228 g/mol
Density:1.145 g/mL (25 °C) (lit.)
:GHS07
Storage Temperature:Ambient
CAS Number:1678-52-0
Formula:C₁₁H₁₃NO₄
MDL Number:MFCD00012349