Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Malachite green solutions
Malachite green solution, Sigma-Aldrich®
Malachite green solution, Sigma-Aldrich®
Malachite green solution, Sigma-Aldrich®
Catalog # HT8028-250ML
Supplier:
SIGMA ALDRICH MICROSCOPY
CAS Number:
Certificates
Expand image
Malachite green solution, Sigma-Aldrich®
Catalog # HT8028-250ML
Supplier:
SIGMA ALDRICH MICROSCOPY
CAS Number:
Certificates
Specifications
Pk:
250 mL
Seq:
2590
Specifications
About this item
Product Details
Documentation
:
ADR:
8,III
:
GHS07
Storage Temperature:
Ambient
:
1
Product Details
Documentation