

3-Indolyl-ß-D-galactopyranoside, Sigma-Aldrich®
Catalog # I7513-5MG
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pk:5 mg
- Seq:2210
Specifications
About this item
:
:295.291 g/mol
ADR:4.1,II
:M-Clarity™ quality level = MQ200
:GHS02
:GHS07
Storage Temperature:Freezer
CAS Number:126787-65-3
Formula:C₁₄H₁₇NO₆
MDL Number:MFCD00083389