Wright's stain modified, Sigma-Aldrich®
Catalog # WS16-500ML
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pk:500 mL
- Seq:2725
- Environmentally Preferable:
Specifications
About this item
Synonyms: Eosin Methylene blue solution according to Wright, Wright's eosin methylene blue
:
ADR:3,III
:GHS02
:GHS06
:GHS08
Storage Temperature:Ambient
:1
CAS Number:68988-92-1
:51938-72-8
MDL Number:MFCD00082143