


Wright's stain modified, Sigma-Aldrich®
Catalog # WS32-1L
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pk:1 L
- Seq:5037
Specifications
About this item
:
ADR:3,III
:GHS02
:GHS06
:GHS08
Storage Temperature:Ambient
:1
CAS Number:68988-92-1
:51938-72-8
MDL Number:MFCD00082143