Bis(cyclohexanone)oxalyldihydrazone, Sigma-Aldrich®
Catalog # C9012-25G
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:278.36 g/mol
Melting Pt:210…214 °C (lit.)
Storage Temperature:Refrigerator
CAS Number:370-81-0
Formula:C₁₄H₂₂N₄O₂
MDL Number:MFCD00001659