Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Bromphenol Blue–Xylene Cyanole
Bromphenol Blue–Xylene Cyanole solution, Sigma-Aldrich®
Bromphenol Blue–Xylene Cyanole solution, Sigma-Aldrich®
Bromphenol Blue–Xylene Cyanole solution, Sigma-Aldrich®
Catalog # B3269-5ML
Supplier:
SIGMA ALDRICH MICROSCOPY
CAS Number:
Expand image
Bromphenol Blue–Xylene Cyanole solution, Sigma-Aldrich®
Catalog # B3269-5ML
Supplier:
SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
Pk:
5 mL
Seq:
2210
Specifications
About this item
Product Details
:
:
GHS08
:
GHS07
Storage Temperature:
Ambient
Product Details