Order Entry
Austria
ContactUsLinkComponent
 
Corrosive
Iron hematoxylin solution (Weigert) Part B, Sigma-Aldrich®
Catalog # HT109-500ML
CAS Number:  
Corrosive
Iron hematoxylin solution (Weigert) Part B, Sigma-Aldrich®
Catalog # HT109-500ML
CAS Number:  

Specifications

  • Pk:
    500 mL
  • Seq:
    2725

Specifications

About this item

 :
ADR:8,III
 :GHS05
Storage Temperature:Ambient
 :1
MDL Number:MFCD00284879