Order Entry
Austria
ContactUsLinkComponent
Giemsa's stain modified, Sigma-Aldrich®
Flammable
Toxic
Health
Giemsa's stain modified, Sigma-Aldrich®
Catalog # GS500-500ML
CAS Number:  
Flammable
Toxic
Health
Giemsa's stain modified, Sigma-Aldrich®
Catalog # GS500-500ML
CAS Number:  

Specifications

  • Pk:
    500 mL
  • Seq:
    2725

Specifications

About this item

 :
ADR:3,II
 :GHS02
 :GHS06
 :GHS08
Storage Temperature:Ambient
 :1
CAS Number:51811-82-6
MDL Number:MFCD00081642