Order Entry
Austria
ContactUsLinkComponent
Sodium nitrite solution, Sigma-Aldrich®
Sodium nitrite solution, Sigma-Aldrich®
Catalog # 914-10ML
CAS Number:  
Sodium nitrite solution, Sigma-Aldrich®
Catalog # 914-10ML
CAS Number:  

Specifications

  • Pk:
    10 mL
  • Seq:
    2275

Specifications

About this item

 :
 :69 g/mol
Storage Temperature:Refrigerator
 :1
CAS Number:7632-00-0
Formula:NaNO₂
MDL Number:MFCD00011118