Sodium fluoride solution, Sigma-Aldrich®
Product Details & Documents
For use in inhibiting α-naphthyl acetate reaction on monocytes. Optional reagent in Sigma Esterase Procedure 91A.
- CAS Number:7681-49-4
- Formula:NaF
- MDL Number:MFCD00003524
- Molecular Weight:41.99 g/mol
- Density:1.005 g/mL (25 °C) (lit.)
- Storage Temperature:Ambient
Documents
Merck The Color of Tissue Diagnostics
Product Literature
Specifications
Specifications
- Pk:25 mL
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



