Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Histo/Zyme

Histo/Zyme
Certificates

2 Options Available Below

Product Details & Documents

Histo/Zyme is a stabilized proteolytic enzyme in buffered solution, used to unmask immunoreactive sites altered by fixation and/or the embedding process. Treatment with a proteolytic enzyme in immunohistochemistry (IHC) is required to digest excess linkages and expose the antigen. The use of Histo/Zyme results in superior staining over traditionally used enzyme pretreatments, such as pepsin and trypsin. Moreover, Histo/Zyme requires only a brief incubation at room temperature. Performing antigen recovery with Histo/Zyme will allow you to dilute the primary antibody 5 to 10 fold further and achieve optimal staining.

  • Antigen Retriever
Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • H3292-15ML
  • H3292-50ML
  • Enzyme name:
  • Histo/Zyme
  • Histo/Zyme
  • Concentration:
  • Ready to Use
  • Ready to Use
  • Storage conditions:
  • 2…8 °C
  • 2…8 °C
  • pH:
  • 7.2
  • 7.2
  • M-Clarity quality level:
  • MQ100
  • MQ100

Specifications

Product Family Options

Product Information

  • Pk
    Availability
    Price
  • 15 mL
    Log in for availability
    Log in for availability
    1 * 15 mL
    1 * 15 mL - 249,00 €
    249,00 €
    1 * 15 mL
       249,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    H3292-15ML
    Enzyme name
    Histo/Zyme
    Concentration
    Ready to Use
    Storage conditions
    2…8 °C
    pH
    7.2
    M-Clarity quality level
    MQ100
  • 50 mL
    Log in for availability
    Log in for availability
    1 * 50 mL
    1 * 50 mL - 694,00 €
    694,00 €
    1 * 50 mL
       694,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    H3292-50ML
    Enzyme name
    Histo/Zyme
    Concentration
    Ready to Use
    Storage conditions
    2…8 °C
    pH
    7.2
    M-Clarity quality level
    MQ100