ContactUsLinkComponent
Disinfectant detergent, Suma Bac D10
Catalog # DIVY100881554
Supplier:  Diversey
Disinfectant detergent, Suma Bac D10
Catalog # DIVY100881554
Supplier:  Diversey
Specifications
  • Description:
    Suma Bac D10 disinfectant detergent
  • Packed:
    6×2 L bottle
  • Pk:
    6 Bottle
Specifications
About this item
Suma Bac D10 is a coloured and unscented bactericidal disinfectant degreaser, which allows the cleaning and disinfection of all surfaces in the food environment in a single operation.
  • Allows cleaning and disinfection in a single operation
  • Attacks grease and soiling generally encountered in food environments
  • Effective on a wide spectrum of microorganisms, improving the level of hygiene
  • Compatible for use with all hardnesses water
It is suitable for cleaning and disinfection in one step on all surfaces generally encountered in the food industry. Its formula, composed in particular of quaternary ammoniums and sequestering agents, makes it particularly effective on a wide spectrum of microorganisms, whatever the hardness of the water. The formula also contains specific surfactants that enhance its cleaning efficiency.
Caution: Use biocides safely. Always read the label and product information before use.